Assay Details
Target Gene Details
Entrez Gene ID: | 3646 |
Gene Name: | eukaryotic translation initiation factor 3 subunit E |
Gene Aliases: |
EIF3-P48, EIF3S6, INT6, eIF3-p46 |
Location: |
Chr.8:108201743-108248802 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EIF3E | NM_001568.2 | NP_001559.1 | ||
XM_017013398.1 | XP_016868887.1 | |||
AK124178.1 | ||||
AK313186.1 | ||||
BC000734.2 | AAH00734.1 | |||
BC008419.1 | AAH08419.1 | |||
BC016706.1 | AAH16706.1 | |||
BC017887.1 | AAH17887.1 | |||
BC021679.1 | AAH21679.1 | |||
CR457029.1 | CAG33310.1 | |||
CR542275.1 | CAG47071.1 | |||
U54562.1 | AAC51760.1 | |||
U62962.1 | AAB58251.1 | |||
U85947.1 | AAB88873.1 | |||
U94175.1 | AAC51919.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |