Assay Details
Target Gene Details
Entrez Gene ID: | 9821 |
Gene Name: | RB1 inducible coiled-coil 1 |
Gene Aliases: |
ATG17, CC1, FIP200, PPP1R131 |
Location: |
Chr.8:52622458-52714466 on Build GRCh38 |
Assay Gene Location: | Within Intron 25 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv611355 | Chr.8:52490251 - 52776552 on Build GRCh38 | Gain | LOC107986877 FAM150A RB1CC1 |
dgv7218n100 | Chr.8:52389530 - 53007050 on Build GRCh38 | Gain | LOC107986877 FAM150A RB1CC1 ST18 NPBWR1 |
esv3891395 | Chr.8:52330215 - 52674146 on Build GRCh38 | Gain | LOC107986877 FAM150A RB1CC1 ST18 |
esv3576572 | Chr.8:52429401 - 52997384 on Build GRCh38 | Gain | LOC107986877 FAM150A RB1CC1 NPBWR1 |
nsv831318 | Chr.8:52626039 - 52811127 on Build GRCh38 | Loss | RB1CC1 |
esv3891396 | Chr.8:52581711 - 52665028 on Build GRCh38 | Gain | RB1CC1 |
esv3617214 | Chr.8:52513334 - 52648974 on Build GRCh38 | Gain | LOC107986877 FAM150A RB1CC1 |
esv2764065 | Chr.8:52452089 - 52975372 on Build GRCh38 | Gain | LOC107986877 FAM150A RB1CC1 NPBWR1 |
esv3617217 | Chr.8:52625360 - 52684547 on Build GRCh38 | Gain | RB1CC1 |
esv2761192 | Chr.8:52365749 - 52700691 on Build GRCh38 | Gain | LOC107986877 FAM150A RB1CC1 ST18 |
dgv7220n100 | Chr.8:52580301 - 52810024 on Build GRCh38 | Gain | RB1CC1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |