Assay Details
Target Gene Details
Entrez Gene ID: | 137970 |
Gene Name: | unc-5 netrin receptor D |
Gene Aliases: |
PRO34692, Unc5h4 |
Location: |
Chr.8:35235457-35796550 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UNC5D | NM_080872.3 | 1 | 243 | NP_543148.2 |
XM_011544400.2 | 1 | 217 | XP_011542702.1 | |
XM_011544401.2 | 1 | 217 | XP_011542703.1 | |
XM_011544402.2 | 1 | 217 | XP_011542704.1 | |
XM_011544403.2 | 1 | 217 | XP_011542705.1 | |
XM_011544404.2 | 1 | 217 | XP_011542706.1 | |
XM_017013044.1 | 1 | 218 | XP_016868533.1 | |
XM_017013045.1 | 1 | 218 | XP_016868534.1 | |
XM_017013046.1 | 1 | 218 | XP_016868535.1 | |
XM_017013047.1 | 1 | 217 | XP_016868536.1 | |
AY358147.1 | 1 | 243 | AAQ88514.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831284 | Chr.8:35160901 - 35335566 on Build GRCh38 | Gain | UNC5D |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |