Assay Details
Target Gene Details
Entrez Gene ID: | 286046 |
Gene Name: | XK related 6 |
Gene Aliases: |
C8orf21, C8orf5, C8orf7, XRG6 |
Location: |
Chr.8:10896144-11205789 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
XKR6 | NM_173683.3 | NP_775954.2 | ||
AY534244.1 | AAT07093.1 | |||
BC024146.2 | AAH24146.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831232 | Chr.8:10748543 - 10917438 on Build GRCh38 | Loss | LOC101929229 XKR6 MIR1322 LOC102723313 LOC105379239 PINX1 |
dgv7011n100 | Chr.8:10734291 - 10946733 on Build GRCh38 | Gain | LOC101929229 XKR6 MIR1322 LOC102723313 LOC105379239 PINX1 |