Assay Details
Target Gene Details
Entrez Gene ID: | 28991 |
Gene Name: | COMM domain containing 5 |
Gene Aliases: |
HCARG, HT002 |
Location: |
Chr.8:144837978-144853578 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
COMMD5 | NM_001081003.2 | NP_001074472.1 | ||
NM_001081004.2 | NP_001074473.1 | |||
NM_001287237.1 | NP_001274166.1 | |||
NM_014066.3 | NP_054785.2 | |||
AF113540.1 | AAF14877.1 | |||
AF290195.1 | AAG09915.1 | |||
AK001477.1 | BAA91714.1 | |||
AK023070.1 | BAB14389.1 | |||
BC002672.2 | AAH02672.1 | |||
BC003055.1 | AAH03055.1 | |||
BC065729.1 | AAH65729.1 | |||
BG705627.1 | ||||
BI753202.1 | ||||
BM013651.1 | ||||
BM924244.1 | ||||
BU508224.1 | ||||
H92104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1023752 | Chr.8:144835600 - 144866312 on Build GRCh38 | Gain | COMMD5 ZNF7 |
nsv469809 | Chr.8:144842323 - 145040108 on Build GRCh38 | Loss | COMMD5 ZNF7 TMED10P1 ZNF252P ZNF250 ZNF16 ZNF252P-AS1 LOC107986987 |
nsv1031737 | Chr.8:144830982 - 144986209 on Build GRCh38 | Gain | COMMD5 ZNF7 ZNF252P ZNF250 ZNF16 |
nsv612969 | Chr.8:144785726 - 144877525 on Build GRCh38 | Loss | COMMD5 ZNF7 ZNF250 ZNF517 ZNF34 MIR6850 RPL8 |
nsv6450 | Chr.8:144850126 - 144882473 on Build GRCh38 | Insertion | COMMD5 ZNF250 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |