Assay Details
Target Gene Details
Entrez Gene ID: | 6575 |
Gene Name: | solute carrier family 20 member 2 |
Gene Aliases: |
GLVR-2, GLVR2, IBGC1, IBGC3, MLVAR, PIT-2, PIT2, RAM1, Ram-1 |
Location: |
Chr.8:42416462-42542213 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC20A2 | NM_006749.4 | 1 | 93 | NP_006740.1 |
XM_017013748.1 | XP_016869237.1 | |||
XM_017013752.1 | XP_016869241.1 | |||
DA418477.1 | 1 | 95 |
Target Gene Details
Entrez Gene ID: | 114926 |
Gene Name: | small integral membrane protein 19 |
Gene Aliases: |
C8orf40 |
Location: |
Chr.8:42541155-42552997 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMIM19 | NM_001135674.1 | 1 | 500 | NP_001129146.1 |
NM_001135675.1 | NP_001129147.1 | |||
NM_001135676.1 | NP_001129148.1 | |||
NM_138436.3 | NP_612445.2 | |||
XM_005273398.4 | 1 | 322 | XP_005273455.1 | |
AK311932.1 | ||||
BC013035.2 | AAH13035.1 | |||
BI335341.1 | 1 | 291 | ||
BI463786.1 | ||||
BI549696.1 | ||||
DA702519.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1017899 | Chr.8:42510362 - 42586679 on Build GRCh38 | Gain | SLC20A2 SMIM19 |
More Information
Additional Information:
For this assay, SNP(s) [rs113755761] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic DGV Variation |