Assay Details
Target Gene Details
Entrez Gene ID: | 252969 |
Gene Name: | nei like DNA glycosylase 2 |
Gene Aliases: |
NEH2, NEI2 |
Location: |
Chr.8:11769663-11787346 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NEIL2 | NM_001135746.1 | NP_001129218.1 | ||
NM_001135747.1 | NP_001129219.1 | |||
NM_001135748.1 | NP_001129220.1 | |||
NM_145043.2 | NP_659480.1 | |||
XM_005272382.2 | XP_005272439.1 | |||
XM_005272383.2 | XP_005272440.1 | |||
XM_017013301.1 | XP_016868790.1 | |||
AB079070.1 | BAC06478.1 | |||
AK056206.1 | BAB71120.1 | |||
AK097389.1 | BAC05030.1 | |||
AK225837.1 | ||||
AK294224.1 | ||||
BC013952.2 | AAH13952.1 | |||
BC013964.2 | AAH13964.1 | |||
BX537529.1 | CAD97774.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3572513 | Chr.8:11776037 - 11784285 on Build GRCh38 | Loss | NEIL2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |