Assay Details
Target Gene Details
Entrez Gene ID: | 665 |
Gene Name: | BCL2 interacting protein 3 like |
Gene Aliases: |
BNIP3a, NIX |
Location: |
Chr.8:26383007-26413128 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BNIP3L | NM_004331.2 | NP_004322.1 | ||
AB004788.1 | BAA28692.1 | |||
AF060922.1 | AAG43134.1 | |||
AF067396.1 | AAD03589.1 | |||
AF079221.1 | AAC27723.1 | |||
AF452712.2 | AAL50978.1 | |||
AF536326.1 | AAN04051.1 | |||
AK290213.1 | ||||
AK300286.1 | ||||
AK300727.1 | ||||
AK307195.1 | ||||
AK315870.1 | ||||
AL132665.1 | CAI46217.1 | |||
BC001559.1 | AAH01559.1 | |||
BC009603.1 | AAH09603.1 | |||
BT019500.1 | AAV38307.1 | |||
BT019501.1 | AAV38308.1 | |||
CD558918.1 | ||||
CR456701.1 | CAG32982.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3572648 | Chr.8:26367602 - 26402306 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv831273 | Chr.8:26358989 - 26513275 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)