Assay Details
Target Gene Details
Entrez Gene ID: | 105375938 |
Gene Name: | translation initiation factor IF-2-like |
Gene Aliases: |
- |
Location: |
Chr.8:60909471-60910926 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105375938 | XM_011517657.2 | XP_011515959.1 | ||
XM_017014120.1 | XP_016869609.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv526196 | Chr.8:60681010 - 60965064 on Build GRCh38 | Gain | CHD7 LOC105375938 |