Assay Details
Target Gene Details
Entrez Gene ID: | 51312 |
Gene Name: | solute carrier family 25 member 37 |
Gene Aliases: |
HT015, MFRN, MFRN1, MSC, MSCP, PRO1278, PRO1584, PRO2217 |
Location: |
Chr.8:23528795-23575463 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC25A37 | NM_001317812.1 | NP_001304741.1 | ||
NM_001317813.1 | NP_001304742.1 | |||
NM_001317814.1 | NP_001304743.1 | |||
NM_016612.3 | NP_057696.2 | |||
XM_011544550.2 | XP_011542852.1 | |||
XM_011544553.2 | XP_011542855.1 | |||
XM_011544554.2 | XP_011542856.1 | |||
XM_017013532.1 | XP_016869021.1 | |||
AF223466.1 | AAF64141.1 | |||
AK074708.1 | ||||
AK075313.1 | ||||
AK127666.1 | ||||
AK223194.1 | BAD96914.1 | |||
AL833186.1 | CAH10415.1 | |||
AY032628.1 | AAK38154.1 | |||
BC015013.2 | AAH15013.1 | |||
BC132799.1 | ||||
BC132801.1 | ||||
BI460337.1 | ||||
BM552840.1 | ||||
BM926580.1 | ||||
BU166030.1 | ||||
DB057758.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508499 | Chr.8:23452508 - 23565200 on Build GRCh38 | Deletion |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)