Assay Details
Target Gene Details
Entrez Gene ID: | 157506 |
Gene Name: | retinol dehydrogenase 10 (all-trans) |
Gene Aliases: |
SDR16C4 |
Location: |
Chr.8:73294602-73325285 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RDH10 | NM_172037.4 | NP_742034.1 | ||
AF456765.1 | AAN64747.1 | |||
AY358270.1 | AAQ88637.1 | |||
BC067131.1 | AAH67131.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2737154 | Chr.8:71825978 - 73469084 on Build GRCh38 | Deletion | RDH10 MSC C8orf89 MSC-AS1 LOC392232 TERF1 LOC101926908 STAU2-AS1 RPL7 KCNB2 TRPA1 RDH10-AS1 STAU2 SBSPON |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |