Assay Details
Target Gene Details
Entrez Gene ID: | 6386 |
Gene Name: | syndecan binding protein |
Gene Aliases: |
MDA-9, MDA9, ST1, SYCL, TACIP18 |
Location: |
Chr.8:58553169-58582860 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SDCBP | NM_001007067.1 | NP_001007068.1 | ||
NM_001007068.1 | NP_001007069.1 | |||
NM_001007069.1 | NP_001007070.1 | |||
NM_001007070.1 | NP_001007071.1 | |||
NM_005625.3 | NP_005616.2 | |||
AF000652.1 | AAB97144.1 | |||
AF006636.1 | AAC52050.1 | |||
AK128645.1 | ||||
AK295198.1 | ||||
AK298697.1 | ||||
AK300647.1 | ||||
AK312274.1 | ||||
BC113674.1 | ||||
BC113676.1 | ||||
BC143915.1 | ||||
BC143916.1 | ||||
BG772269.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)