Assay Details
Target Gene Details
Entrez Gene ID: | 100130742 |
Gene Name: | leucine rich repeat containing 69 |
Gene Aliases: |
- |
Location: |
Chr.8:91102619-91219236 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRRC69 | NM_001129890.1 | NP_001123362.1 | ||
AK097393.1 | ||||
AK130865.1 | BAC85448.1 |
Target Gene Details
Entrez Gene ID: | 115111 |
Gene Name: | solute carrier family 26 member 7 |
Gene Aliases: |
SUT2 |
Location: |
Chr.8:91209494-91398155 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC26A7 | NM_001282356.1 | NP_001269285.1 | ||
NM_001282357.1 | NP_001269286.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759629 | Chr.8:90962271 - 91261496 on Build GRCh38 | Loss | MIR4661 OTUD6B LRRC69 OTUD6B-AS1 C8orf88 SLC26A7 TMEM55A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |