Assay Details
Target Gene Details
Entrez Gene ID: | 64478 |
Gene Name: | CUB and Sushi multiple domains 1 |
Gene Aliases: |
PPP1R24 |
Location: |
Chr.8:2935353-4995035 on Build GRCh38 |
Assay Gene Location: | Within Intron 71 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CSMD1 | NM_033225.5 | NP_150094.5 | ||
XM_011534752.2 | XP_011533054.1 | |||
XM_011534753.2 | XP_011533055.1 | |||
XM_011534754.1 | XP_011533056.1 | |||
AB209502.1 | BAD92739.1 | |||
AF333704.2 | AAK73475.2 | |||
AK127129.1 | ||||
AY017307.1 | AAG52948.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1033589 | Chr.8:2490387 - 3447578 on Build GRCh38 | Loss | LOC105377792 CSMD1 LOC101927815 |
dgv1357e214 | Chr.8:2895602 - 3065852 on Build GRCh38 | Gain | LOC105377792 CSMD1 |
esv3584570 | Chr.8:2827824 - 3129358 on Build GRCh38 | Gain | LOC105377792 CSMD1 |
esv3891261 | Chr.8:2316229 - 3462226 on Build GRCh38 | Gain | LOC105377792 CSMD1 LOC101927815 |
esv3615892 | Chr.8:2811468 - 2947245 on Build GRCh38 | Gain | CSMD1 |
esv3572405 | Chr.8:2939846 - 2947404 on Build GRCh38 | Loss | CSMD1 |
dgv6815n100 | Chr.8:2193919 - 3295843 on Build GRCh38 | Gain | LOC105377792 CSMD1 LOC105377781 LOC101927815 |
nsv609710 | Chr.8:2872547 - 3175339 on Build GRCh38 | Gain | LOC105377792 CSMD1 |
dgv11848n54 | Chr.8:2320544 - 3445528 on Build GRCh38 | Gain | LOC105377792 CSMD1 LOC101927815 |
esv3584572 | Chr.8:2935217 - 3102350 on Build GRCh38 | Gain | LOC105377792 CSMD1 |
nsv1027569 | Chr.8:2660367 - 3086961 on Build GRCh38 | Loss | LOC105377792 CSMD1 LOC101927815 |
nsv1024325 | Chr.8:651043 - 3203837 on Build GRCh38 | Gain | LOC105377776 KBTBD11 LOC101927752 MIR7160 DLGAP2-AS1 LOC101927815 MIR596 KBTBD11-OT1 ERICH1-AS1 DLGAP2 CLN8 LOC401442 LOC101928058 LOC105377792 LOC100131395 ARHGEF10 LOC105377774 MIR3674 ERICH1 CSMD1 MYOM2 LOC105377781 LOC286083 LOC105377777 |
nsv517808 | Chr.8:2269008 - 3111255 on Build GRCh38 | Gain | LOC105377792 CSMD1 LOC101927815 |
nsv1034294 | Chr.8:2901778 - 3054246 on Build GRCh38 | Gain | LOC105377792 CSMD1 |
nsv1018407 | Chr.8:2866772 - 3447578 on Build GRCh38 | Gain | LOC105377792 CSMD1 |
nsv831209 | Chr.8:2767402 - 2954003 on Build GRCh38 | Loss | CSMD1 |
dgv6827n100 | Chr.8:2473547 - 3062661 on Build GRCh38 | Gain | LOC105377792 CSMD1 LOC101927815 |
More Information
Additional Information:
For this assay, SNP(s) [rs78111778] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |