Assay Details
Target Gene Details
Entrez Gene ID: | 25879 |
Gene Name: | DDB1 and CUL4 associated factor 13 |
Gene Aliases: |
GM83, HSPC064, WDSOF1 |
Location: |
Chr.8:103414714-103443453 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DCAF13 | NM_015420.6 | NP_056235.4 | ||
AF161549.1 | AAF29036.1 | |||
AK001693.1 | ||||
AK001874.1 | BAA91955.1 | |||
AK027799.1 | BAB55377.1 | |||
AK074725.1 | BAC11163.1 | |||
BC026067.1 | AAH26067.2 | |||
BC101810.1 | AAI01811.1 | |||
BC112042.1 | AAI12043.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv611832 | Chr.8:103388580 - 103469507 on Build GRCh38 | Loss |
![]() ![]() |
dgv7279n100 | Chr.8:103349626 - 103867456 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)