Assay Details
Target Gene Details
Entrez Gene ID: | 84154 |
Gene Name: | ribosome production factor 2 homolog |
Gene Aliases: |
BXDC1, bA397G5.4 |
Location: |
Chr.6:110982017-111028263 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RPF2 | NM_001289111.1 | NP_001276040.1 | ||
NM_032194.2 | NP_115570.1 | |||
AK024740.1 | BAB14983.1 | |||
AK292165.1 | ||||
AK301662.1 | ||||
AL833613.1 | ||||
BC035314.1 | AAH35314.1 | |||
BG529605.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1032572 | Chr.6:110800096 - 111060820 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv1020627 | Chr.6:110982539 - 111302511 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv1016001 | Chr.6:110959208 - 110988298 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)