Assay Details
Target Gene Details
Entrez Gene ID: | 51106 |
Gene Name: | transcription factor B1, mitochondrial |
Gene Aliases: |
CGI-75, CGI75, mtTFB, mtTFB1 |
Location: |
Chr.6:155229990-155314497 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TFB1M |
Target Gene Details
Entrez Gene ID: | 26230 |
Gene Name: | T-cell lymphoma invasion and metastasis 2 |
Gene Aliases: |
STEF, TIAM-2 |
Location: |
Chr.6:155090289-155257723 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TIAM2 | NM_001010927.2 | NP_001010927.1 | ||
NM_012454.3 | NP_036586.2 | |||
AF120323.1 | AAF05900.1 | |||
AF120324.1 | AAF05901.1 | |||
AK123859.1 | ||||
AK125459.1 | BAC86170.1 | |||
AK299723.1 | ||||
AK302860.1 | ||||
AL122086.1 | CAB59259.2 | |||
BC066979.1 | AAH66979.1 | |||
BC137275.1 | ||||
BP199501.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv526576 | Chr.6:155230087 - 155258266 on Build GRCh38 | Loss | TFB1M TIAM2 |
nsv1025003 | Chr.6:155077125 - 155755140 on Build GRCh38 | Gain | NOX3 TFB1M LOC105378068 TIAM2 CLDN20 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |