Assay Details
Target Gene Details
Entrez Gene ID: | 64065 |
Gene Name: | PERP, TP53 apoptosis effector |
Gene Aliases: |
KCP1, KRTCAP1, PIGPC1, THW, dJ496H19.1 |
Location: |
Chr.6:138088505-138107523 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PERP | NM_022121.4 | NP_071404.2 | ||
AF317550.1 | AAG35063.1 | |||
AJ251830.1 | CAC17766.1 | |||
AK074585.1 | BAC11074.1 | |||
AK075082.1 | BAC11390.1 | |||
AK093516.1 | ||||
AK097387.1 | ||||
AK097958.1 | BAC05205.1 | |||
AK225907.1 | ||||
AK314526.1 | ||||
AY157578.1 | AAO13162.1 | |||
BC010163.2 | AAH10163.1 | |||
CX784592.1 | ||||
DA750046.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |