Assay Details
Target Gene Details
Entrez Gene ID: | 221711 |
Gene Name: | synaptonemal complex protein 2 like |
Gene Aliases: |
C6orf177, NO145, dJ62D2.1 |
Location: |
Chr.6:10886831-10974309 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SYCP2L | NM_001040274.2 | NP_001035364.2 | ||
AK128130.1 | BAC87288.1 | |||
AM293674.1 | ||||
DB077181.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv519302 | Chr.6:10879763 - 10892266 on Build GRCh38 | Loss | GCM2 SYCP2L |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |