Assay Details
Target Gene Details
Entrez Gene ID: | 55510 |
Gene Name: | DEAD-box helicase 43 |
Gene Aliases: |
CT13, HAGE |
Location: |
Chr.6:73394562-73417566 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDX43 | NM_018665.2 | NP_061135.2 | ||
XM_011535926.1 | XP_011534228.1 | |||
XM_011535928.2 | XP_011534230.1 | |||
XM_017011003.1 | XP_016866492.1 | |||
AJ278110.1 | CAB92442.1 | |||
AK303324.1 | ||||
AK313686.1 | ||||
AL136751.1 | CAB66685.1 | |||
BC066938.1 | AAH66938.1 | |||
BX646425.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830693 | Chr.6:73339619 - 73506357 on Build GRCh38 | Loss | MTO1 MB21D1 OOEP DPPA5 KHDC3L DDX43 OOEP-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |