Assay Details
Target Gene Details
Entrez Gene ID: | 4552 |
Gene Name: | 5-methyltetrahydrofolate-homocysteine methyltransferase reductase |
Gene Aliases: |
MSR, cblE |
Location: |
Chr.5:7869104-7901124 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MTRR | NM_002454.2 | NP_002445.2 | ||
NM_024010.2 | NP_076915.2 | |||
NR_134480.1 | ||||
NR_134481.1 | ||||
NR_134482.1 | ||||
AF025794.1 | AAC39667.1 | |||
AF121214.1 | AAF16876.1 | |||
AK308442.1 | ||||
AK311663.1 | ||||
BC054816.1 | ||||
BC109216.1 | AAI09217.1 | |||
DA384356.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv597005 | Chr.5:7860291 - 8092964 on Build GRCh38 | Gain | MTRR FASTKD3 |
nsv597003 | Chr.5:7538124 - 8706231 on Build GRCh38 | Gain | ADCY2 MIR4458HG MIR4458 MTRR C5orf49 FASTKD3 LOC729506 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |