Assay Details
Target Gene Details
Entrez Gene ID: | 1879 |
Gene Name: | early B-cell factor 1 |
Gene Aliases: |
COE1, EBF, O/E-1, OLF1 |
Location: |
Chr.5:158695915-159099762 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EBF1 | NM_001290360.2 | NP_001277289.1 | ||
NM_001324101.1 | NP_001311030.1 | |||
NM_001324103.1 | NP_001311032.1 | |||
NM_001324106.1 | NP_001311035.1 | |||
NM_001324107.1 | NP_001311036.1 | |||
NM_001324108.1 | NP_001311037.1 | |||
NM_001324109.1 | NP_001311038.1 | |||
NM_001324111.1 | NP_001311040.1 | |||
NM_024007.4 | NP_076870.1 | |||
NM_182708.2 | NP_874367.1 | |||
XM_017009198.1 | XP_016864687.1 | |||
XM_017009201.1 | XP_016864690.1 | |||
AF208502.1 | AAF19643.1 | |||
AK289712.1 | ||||
AK303698.1 | ||||
AK304438.1 | ||||
BC038805.1 | AAH38805.1 | |||
BC041178.1 | AAH41178.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv5093 | Chr.5:158691822 - 158725743 on Build GRCh38 | Insertion | EBF1 |
nsv830528 | Chr.5:158698214 - 158913021 on Build GRCh38 | Gain | EBF1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |