Assay Details
Target Gene Details
Entrez Gene ID: | 3003 |
Gene Name: | granzyme K |
Gene Aliases: |
TRYP2 |
Location: |
Chr.5:55024279-55034132 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GZMK | NM_002104.2 | NP_002095.1 | ||
AK312074.1 | ||||
BC035802.1 | AAH35802.1 | |||
U26174.1 | AAA74578.1 | |||
U35237.1 | AAA79063.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830301 | Chr.5:54930046 - 55127677 on Build GRCh38 | Gain | ESM1 GZMA LOC102467081 CDC20B GZMK |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |