Assay Details
Target Gene Details
Entrez Gene ID: | 3074 |
Gene Name: | hexosaminidase subunit beta |
Gene Aliases: |
ENC-1AS, HEL-248, HEL-S-111 |
Location: |
Chr.5:74640023-74721288 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HEXB | NM_001292004.1 | NP_001278933.1 | ||
AY643499.1 | AAV48875.1 | |||
FJ224336.1 | ||||
GQ472222.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |