Assay Details
Target Gene Details
Entrez Gene ID: | 133383 |
Gene Name: | SET domain containing 9 |
Gene Aliases: |
C5orf35 |
Location: |
Chr.5:56909260-56928900 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SETD9 | NM_001171990.2 | NP_001165461.1 | ||
NR_136555.1 | ||||
AK309152.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv598210 | Chr.5:56881616 - 56958658 on Build GRCh38 | Loss | SETD9 MAP3K1 MIER3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |