Assay Details
Target Gene Details
Entrez Gene ID: | 10007 |
Gene Name: | glucosamine-6-phosphate deaminase 1 |
Gene Aliases: |
GNP1, GNPDA, GNPI, GPI, HLN |
Location: |
Chr.5:142000669-142013055 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GNPDA1 | NM_005471.4 | NP_005462.1 | ||
XM_005268348.1 | XP_005268405.1 | |||
XM_006714747.2 | XP_006714810.1 | |||
XM_011537537.1 | XP_011535839.1 | |||
AF029914.1 | AAB84217.1 | |||
AF048826.1 | AAC05123.1 | |||
AJ002231.1 | CAA05259.1 | |||
AK022779.1 | ||||
AK290686.1 | ||||
AK296452.1 | ||||
BC012853.2 | AAH12853.1 | |||
BC020769.1 | AAH20769.1 | |||
BC022322.1 | AAH22322.1 | |||
D31766.1 | ||||
DA661926.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1154058 | Chr.5:141632251 - 142199943 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)