Assay Details
Target Gene Details
Entrez Gene ID: | 8817 |
Gene Name: | fibroblast growth factor 18 |
Gene Aliases: |
FGF-18, ZFGF5 |
Location: |
Chr.5:171419663-171457626 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FGF18 | NM_003862.2 | NP_003853.1 | ||
AB007422.1 | BAA31986.1 | |||
AF075292.1 | AAC62240.1 | |||
AF211188.1 | AAF22977.1 | |||
AY358811.1 | AAQ89954.1 | |||
BC006245.2 | AAH06245.1 | |||
BT019570.1 | AAV38377.1 | |||
BT019571.1 | AAV38378.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830551 | Chr.5:171319918 - 171495029 on Build GRCh38 | Loss | FGF18 MIR3912 NPM1 |
nsv830552 | Chr.5:171337559 - 171579435 on Build GRCh38 | Gain | FGF18 MIR3912 NPM1 |
nsv600278 | Chr.5:171418055 - 171471294 on Build GRCh38 | Loss | FGF18 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |