Assay Details
Target Gene Details
Entrez Gene ID: | 54888 |
Gene Name: | NOP2/Sun RNA methyltransferase family member 2 |
Gene Aliases: |
MISU, MRT5, SAKI, TRM4 |
Location: |
Chr.5:6599239-6633360 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NSUN2 | NM_001193455.1 | NP_001180384.1 | ||
NM_017755.5 | NP_060225.4 | |||
NR_037947.1 |
Target Gene Details
Entrez Gene ID: | 6715 |
Gene Name: | steroid 5 alpha-reductase 1 |
Gene Aliases: |
S5AR 1 |
Location: |
Chr.5:6633322-6669562 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SRD5A1 | NM_001047.3 | 1 | 112 | NP_001038.1 |
NM_001324322.1 | 1 | 112 | NP_001311251.1 | |
NM_001324323.1 | 1 | 112 | NP_001311252.1 | |
NR_136739.1 | 1 | 112 | ||
AF052126.1 | AAC28620.1 | |||
AF113128.1 | AAF14869.1 | |||
AK297208.1 | ||||
AK297511.1 | ||||
AK313070.1 | ||||
BC007033.1 | AAH07033.1 | |||
DA170063.1 | ||||
DC350933.1 | 1 | 112 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic non-DGV Variation |