Assay Details
Target Gene Details
Entrez Gene ID: | 133686 |
Gene Name: | NAD kinase 2, mitochondrial |
Gene Aliases: |
C5orf33, DECRD, MNADK, NADKD1 |
Location: |
Chr.5:36192589-36242279 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NADK2 | NM_001085411.2 | NP_001078880.1 | ||
NM_001287340.1 | NP_001274269.1 | |||
NM_001287341.1 | NP_001274270.1 | |||
NM_153013.4 | NP_694558.1 | |||
XM_005248241.4 | XP_005248298.1 | |||
AK055158.1 | BAB70864.1 | |||
AK057950.1 | ||||
AK304009.1 | ||||
AY360463.1 | AAQ62967.1 | |||
BC032912.1 | AAH32912.1 | |||
BC062567.1 | AAH62567.1 | |||
BX648388.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3604707 | Chr.5:35513484 - 36501574 on Build GRCh38 | Gain | NADK2 LOC100506406 LMBRD2 SPEF2 UGT3A1 SKP2 UGT3A2 RANBP3L CAPSL MIR580 IL7R |
nsv830262 | Chr.5:36074209 - 36293770 on Build GRCh38 | Gain | NADK2 LMBRD2 SKP2 RANBP3L MIR580 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |