Assay Details
Target Gene Details
Entrez Gene ID: | 55568 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 10 |
Gene Aliases: |
GALNACT10, PPGALNACT10, PPGANTASE10 |
Location: |
Chr.5:154190735-154420984 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GALNT10 | NM_198321.3 | NP_938080.1 | ||
AB078145.1 | BAC56890.1 | |||
AJ505950.1 | CAD44532.1 | |||
AK074132.1 | BAB84958.1 | |||
AK127135.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |