Assay Details
Target Gene Details
Entrez Gene ID: | 2878 |
Gene Name: | glutathione peroxidase 3 |
Gene Aliases: |
GPx-P, GSHPx-3, GSHPx-P |
Location: |
Chr.5:151020438-151028993 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPX3 | NM_002084.3 | NP_002075.2 | ||
AF217787.1 | AAF43005.1 | |||
AK310242.1 | ||||
BC013601.1 | AAH13601.1 | |||
BC035841.1 | AAH35841.1 | |||
BC050378.1 | AAH50378.2 | |||
BC066949.1 | AAH66949.1 | |||
BI522602.1 | ||||
D00632.1 | BAA00525.1 | |||
X58295.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv526224 | Chr.5:151003841 - 151070675 on Build GRCh38 | Gain | GPX3 TNIP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |