Assay Details
Target Gene Details
Entrez Gene ID: | 26762 |
Gene Name: | hepatitis A virus cellular receptor 1 |
Gene Aliases: |
CD365, HAVCR, HAVCR-1, KIM-1, KIM1, TIM, TIM-1, TIM1, TIMD-1, TIMD1 |
Location: |
Chr.5:156979443-157059119 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HAVCR1 | XM_017009340.1 | XP_016864829.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1016820 | Chr.5:157003694 - 157055302 on Build GRCh38 | Gain | HAVCR1 |
esv3607302 | Chr.5:156839545 - 157055054 on Build GRCh38 | Gain | TIMD4 PPP1R2P3 HAVCR1 |
esv3607306 | Chr.5:156969763 - 157052820 on Build GRCh38 | Gain | HAVCR1 |
esv2730954 | Chr.5:157017808 - 157063108 on Build GRCh38 | Deletion | HAVCR1 |
esv3575892 | Chr.5:156975204 - 157220778 on Build GRCh38 | Gain | HAVCR2 FAM71B ITK HAVCR1 MED7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |