Assay Details
Target Gene Details
Entrez Gene ID: | 6902 |
Gene Name: | tubulin folding cofactor A |
Gene Aliases: |
- |
Location: |
Chr.5:77691169-77776361 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TBCA | NM_001297738.1 | NP_001284667.1 | ||
NM_001297740.1 | NP_001284669.1 | |||
NM_004607.2 | NP_004598.1 | |||
AF038952.1 | AAC39866.1 | |||
AK300031.1 | ||||
AK312207.1 | ||||
BC018210.1 | AAH18210.1 | |||
BC105285.1 | AAI05286.1 | |||
BT007354.1 | AAP36018.1 | |||
CR542170.1 | CAG46967.1 | |||
CR542186.1 | CAG46983.1 | |||
DC312139.1 | ||||
DW436723.1 | ||||
HY015414.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |