Assay Details
Target Gene Details
Entrez Gene ID: | 30820 |
Gene Name: | potassium voltage-gated channel interacting protein 1 |
Gene Aliases: |
KCHIP1, VABP |
Location: |
Chr.5:170353055-170736632 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCNIP1 | NM_001034837.2 | NP_001030009.1 | ||
NM_001034838.2 | NP_001030010.1 | |||
NM_001278339.1 | NP_001265268.1 | |||
NM_001278340.1 | NP_001265269.1 | |||
NM_014592.3 | NP_055407.1 | |||
XM_017009407.1 | XP_016864896.1 | |||
AF199597.1 | AAF33682.1 | |||
AI884645.1 | ||||
AK074390.1 | ||||
AK301775.1 | ||||
AY170821.1 | AAN77491.1 | |||
AY780424.1 | AAV51968.1 | |||
BC050375.1 | AAH50375.1 | |||
BF514334.1 | ||||
DC420716.1 | ||||
DQ148476.1 | AAZ77793.1 | |||
DQ148477.1 | AAZ77794.1 | |||
DQ148478.1 | AAZ77795.1 | |||
DQ148479.1 | AAZ77796.1 |
Target Gene Details
Entrez Gene ID: | 105377716 |
Gene Name: | uncharacterized LOC105377716 |
Gene Aliases: |
- |
Location: |
Chr.5:170639158-170681470 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105377716 | NR_136214.1 | |||
AA626712.1 | ||||
BX116959.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv600262 | Chr.5:170661487 - 171342848 on Build GRCh38 | Gain | TLX3 GABRP KCNIP1 LOC728145 LOC105377716 RANBP17 |
nsv830549 | Chr.5:170569400 - 170724385 on Build GRCh38 | Gain | KCNIP1 LOC105377716 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |