Assay Details
Target Gene Details
Entrez Gene ID: | 2668 |
Gene Name: | glial cell derived neurotrophic factor |
Gene Aliases: |
ATF1, ATF2, HFB1-GDNF, HSCR3 |
Location: |
Chr.5:37812677-37840044 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GDNF | NM_000514.3 | NP_000505.1 | ||
NM_001190468.1 | NP_001177397.1 | |||
NM_001190469.1 | NP_001177398.1 | |||
NM_001278098.1 | NP_001265027.1 | |||
NM_199231.2 | NP_954701.1 | |||
XM_011514030.2 | XP_011512332.1 | |||
XM_017009337.1 | XP_016864826.1 | |||
AF053748.1 | AAD43139.1 | |||
AJ001897.1 | CAA05075.1 | |||
AJ001898.1 | CAA05076.1 | |||
AJ001899.1 | CAA05077.1 | |||
AJ001900.1 | CAA05078.1 | |||
AY052832.1 | AAL11017.1 | |||
BC069119.1 | AAH69119.1 | |||
BC069369.1 | AAH69369.1 | |||
BC128108.1 | ||||
BC128109.1 | ||||
CR541923.1 | CAG46721.1 | |||
DQ235474.1 | ||||
KU178093.1 | ||||
KU178094.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3604748 | Chr.5:37720912 - 37848930 on Build GRCh38 | Gain |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)