Assay Details
Target Gene Details
Entrez Gene ID: | 23138 |
Gene Name: | NEDD4 binding protein 3 |
Gene Aliases: |
LZTS4 |
Location: |
Chr.5:178113506-178127034 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
N4BP3 | NM_015111.1 | NP_055926.1 | ||
XM_006714834.3 | XP_006714897.1 | |||
XM_011534473.1 | XP_011532775.1 | |||
XM_011534474.1 | XP_011532776.1 | |||
BC053323.1 | AAH53323.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1019404 | Chr.5:178080079 - 178432088 on Build GRCh38 | Gain | PHYKPL GMCL1P1 NHP2 N4BP3 COL23A1 RMND5B HNRNPAB |
nsv462567 | Chr.5:178078020 - 178145926 on Build GRCh38 | Loss | N4BP3 RMND5B |
dgv5844n100 | Chr.5:177883324 - 178184647 on Build GRCh38 | Gain | FAM153C GMCL1P1 NHP2 N4BP3 RMND5B PROP1 LOC100288656 LOC100128340 LOC728554 |
nsv600391 | Chr.5:178061280 - 178122490 on Build GRCh38 | Loss | N4BP3 |