Assay Details
Target Gene Details
Entrez Gene ID: | 153328 |
Gene Name: | solute carrier family 25 member 48 |
Gene Aliases: |
- |
Location: |
Chr.5:135579002-135888637 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC25A48 | XM_017009090.1 | XP_016864579.1 | ||
XM_017009092.1 | XP_016864581.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |