Assay Details
Target Gene Details
Entrez Gene ID: | 2246 |
Gene Name: | fibroblast growth factor 1 |
Gene Aliases: |
AFGF, ECGF, ECGF-beta, ECGFA, ECGFB, FGF-1, FGF-alpha, FGFA, GLIO703, HBGF-1, HBGF1 |
Location: |
Chr.5:142592178-142698070 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1034911 | Chr.5:142554309 - 142652186 on Build GRCh38 | Gain | FGF1 |
esv3606999 | Chr.5:142429551 - 143065624 on Build GRCh38 | Gain | ARHGAP26 ARHGAP26-AS1 LOC101926975 LOC101926941 FGF1 |
nsv1031126 | Chr.5:142514198 - 143020006 on Build GRCh38 | Gain | ARHGAP26 ARHGAP26-AS1 LOC101926975 FGF1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |