Assay Details
Target Gene Details
Entrez Gene ID: | 100861518 |
Gene Name: | P4HA2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.5:132184876-132192805 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
P4HA2-AS1 | NR_047470.1 | |||
BG772701.1 |