Assay Details
Target Gene Details
Entrez Gene ID: | 5734 |
Gene Name: | prostaglandin E receptor 4 |
Gene Aliases: |
EP4, EP4R |
Location: |
Chr.5:40679930-40740932 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTGER4 | XM_017009659.1 | XP_016865148.1 |
Target Gene Details
Entrez Gene ID: | 23548 |
Gene Name: | tetratricopeptide repeat domain 33 |
Gene Aliases: |
OSRF |
Location: |
Chr.5:40711576-40756965 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TTC33 | NM_012382.2 | NP_036514.1 | ||
XM_011514003.2 | XP_011512305.1 | |||
AF023244.1 | AAD09341.1 | |||
AK303119.1 | ||||
AK307131.1 | ||||
AK312560.1 | ||||
BC015701.1 | AAH15701.1 | |||
BC036536.1 | AAH36536.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |