Assay Details
Target Gene Details
Entrez Gene ID: | 79685 |
Gene Name: | SAP30 like |
Gene Aliases: |
NS4ATP2 |
Location: |
Chr.5:154445957-154461053 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SAP30L | NM_001131062.1 | NP_001124534.1 | ||
NM_001131063.1 | NP_001124535.1 | |||
NM_024632.5 | NP_078908.1 | |||
NR_024084.1 | ||||
AF085957.1 | ||||
AI199517.1 | ||||
AI436556.1 | ||||
AK021588.1 | BAB13848.1 | |||
AY341060.1 | AAQ16562.1 | |||
AY846876.1 | AAX54477.1 | |||
BC009829.2 | AAH09829.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830520 | Chr.5:154372285 - 154553755 on Build GRCh38 | Loss | HAND1 GALNT10 SAP30L SAP30L-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |