Assay Details
Target Gene Details
Entrez Gene ID: | 345757 |
Gene Name: | family with sequence similarity 174 member A |
Gene Aliases: |
HGS_RE408, TMEM157, UNQ1912 |
Location: |
Chr.5:100535305-100586741 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM174A | NM_198507.2 | NP_940909.1 | ||
XM_006714600.1 | XP_006714663.1 | |||
AF529367.1 | AAQ09601.1 | |||
AK289539.1 | ||||
AY359108.1 | AAQ89466.1 | |||
BC027332.1 | AAH27332.1 | |||
BI462478.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759359 | Chr.5:99871970 - 100547667 on Build GRCh38 | Gain+Loss | FAM174A LOC100133050 |
esv3890678 | Chr.5:100324445 - 101099725 on Build GRCh38 | Gain | ST8SIA4 FAM174A MIR548P LOC100133050 |
nsv599165 | Chr.5:100133300 - 100727335 on Build GRCh38 | Loss | FAM174A LOC100133050 |
nsv1021183 | Chr.5:100290153 - 100885722 on Build GRCh38 | Gain | ST8SIA4 FAM174A MIR548P LOC100133050 |