Assay Details
Target Gene Details
Entrez Gene ID: | 10283 |
Gene Name: | CWC27 spliceosome associated protein homolog |
Gene Aliases: |
NY-CO-10, SDCCAG-10, SDCCAG10 |
Location: |
Chr.5:64768918-65018763 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CWC27 | NM_001297644.1 | NP_001284573.1 | ||
NM_001297645.1 | NP_001284574.1 | |||
NM_001318000.1 | NP_001304929.1 | |||
NM_005869.3 | NP_005860.2 | |||
XM_011543095.2 | XP_011541397.2 | |||
XM_011543096.1 | XP_011541398.1 | |||
XM_017008943.1 | XP_016864432.1 | |||
AF039692.1 | AAC18041.1 | |||
AK225105.1 | ||||
AK225161.1 | ||||
AK308223.1 | ||||
AK312954.1 | ||||
AY358569.1 | AAQ88932.1 | |||
BC012117.1 | AAH12117.1 | |||
BX647971.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1031566 | Chr.5:64737106 - 64787266 on Build GRCh38 | Gain | CWC27 SREK1IP1 |
nsv598390 | Chr.5:64554330 - 64982002 on Build GRCh38 | Gain | RGS7BP CWC27 FAM159B SREK1IP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |