Assay Details
Target Gene Details
Entrez Gene ID: | 65056 |
Gene Name: | GC-rich promoter binding protein 1 |
Gene Aliases: |
GPBP, SSH6, VASCULIN |
Location: |
Chr.5:57173826-57264679 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPBP1 | NM_001127235.2 | NP_001120707.2 | ||
NM_001203246.1 | NP_001190175.1 | |||
NM_022913.3 | NP_075064.1 | |||
XM_006714673.3 | XP_006714736.1 | |||
XM_006714674.3 | XP_006714737.1 | |||
XM_017009754.1 | XP_016865243.1 | |||
XM_017009755.1 | XP_016865244.1 | |||
XM_017009756.1 | XP_016865245.1 | |||
XM_017009757.1 | XP_016865246.1 | |||
XM_017009758.1 | XP_016865247.1 | |||
XM_017009759.1 | XP_016865248.1 | |||
AL161991.1 | CAB82324.2 | |||
AY226828.1 | AAO34124.1 | |||
BC000267.1 | AAH00267.1 | |||
BC113004.2 | AAI13005.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3894192 | Chr.5:57175963 - 57204599 on Build GRCh38 | Loss | GPBP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |