Assay Details
Target Gene Details
Entrez Gene ID: | 3295 |
Gene Name: | hydroxysteroid 17-beta dehydrogenase 4 |
Gene Aliases: |
DBP, MFE-2, MPF-2, PRLTS1, SDR8C1 |
Location: |
Chr.5:119452443-119542335 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HSD17B4 | NM_000414.3 | NP_000405.1 | ||
NM_001199291.2 | NP_001186220.1 | |||
NM_001199292.1 | NP_001186221.1 | |||
NM_001292027.1 | NP_001278956.1 | |||
NM_001292028.1 | NP_001278957.1 | |||
AK094049.1 | ||||
AK293254.1 | ||||
AK298075.1 | ||||
AK299685.1 | ||||
AK301212.1 | ||||
AK312449.1 | ||||
BC003098.1 | AAH03098.1 | |||
DA296575.1 | ||||
DA426062.1 | ||||
DA508630.1 | ||||
DC325873.1 | ||||
KU178135.1 | ||||
KU178136.1 | ||||
KU178137.1 | ||||
KU178138.1 | ||||
X87176.1 | CAA60643.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv4974 | Chr.5:119414564 - 119459681 on Build GRCh38 | Deletion | HSD17B4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |