Assay Details
Target Gene Details
Entrez Gene ID: | 29093 |
Gene Name: | mitochondrial ribosomal protein L22 |
Gene Aliases: |
HSPC158, L22mt, MRP-L22, MRP-L25, RPML25 |
Location: |
Chr.5:154941073-154969411 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MRPL22 | NM_001014990.2 | NP_001014990.1 | ||
NM_014180.3 | NP_054899.2 | |||
AF161507.1 | AAF29122.1 | |||
AK000601.1 | BAA91282.1 | |||
AK302784.1 | ||||
BC012565.1 | AAH12565.1 | |||
BM662504.1 | ||||
BP372420.1 | ||||
BU943770.1 | ||||
CR933679.1 | CAI45973.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)