Assay Details
Target Gene Details
Entrez Gene ID: | 6444 |
Gene Name: | sarcoglycan delta |
Gene Aliases: |
35DAG, CMD1L, DAGD, SG-delta, SGCDP, SGD |
Location: |
Chr.5:155728636-156767788 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SGCD | XM_017009724.1 | XP_016865213.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3607264 | Chr.5:155733910 - 155740456 on Build GRCh38 | Loss | SGCD |
dgv5823n100 | Chr.5:155731480 - 156135824 on Build GRCh38 | Gain | SGCD |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |