Assay Details
Target Gene Details
Entrez Gene ID: | 22948 |
Gene Name: | chaperonin containing TCP1 subunit 5 |
Gene Aliases: |
CCT-epsilon, CCTE, HEL-S-69, PNAS-102, TCP-1-epsilon |
Location: |
Chr.5:10249921-10266412 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCT5 | NM_001306153.1 | NP_001293082.1 | ||
NM_001306154.1 | NP_001293083.1 | |||
NM_001306155.1 | NP_001293084.1 | |||
NM_001306156.1 | NP_001293085.1 | |||
NM_012073.4 | NP_036205.1 | |||
AK289353.1 | ||||
AK290393.1 | ||||
AK293341.1 | ||||
AK293437.1 | ||||
AK301760.1 | ||||
AK301985.1 | ||||
AK302368.1 | ||||
AK302383.1 | ||||
AK303086.1 | ||||
AK303147.1 | ||||
AK316376.1 | ||||
BC002971.1 | AAH02971.1 | |||
BC006543.2 | AAH06543.1 | |||
BC009454.1 | AAH09454.1 | |||
BC035499.2 | AAH35499.1 | |||
D43950.2 | ||||
FJ224303.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv461975 | Chr.5:10236924 - 10262046 on Build GRCh38 | Gain | FAM173B CCT5 |
nsv1029129 | Chr.5:10133592 - 10300387 on Build GRCh38 | Gain | FAM173B CCT5 CMBL |
esv33231 | Chr.5:9985609 - 10272597 on Build GRCh38 | Loss | FAM173B CCT5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |