Assay Details
Target Gene Details
Entrez Gene ID: | 6531 |
Gene Name: | solute carrier family 6 member 3 |
Gene Aliases: |
DAT, DAT1, PKDYS |
Location: |
Chr.5:1392790-1445428 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC6A3 | NM_001044.4 | NP_001035.1 | ||
AK310589.1 | ||||
BC132977.1 | ||||
BC133003.1 | ||||
EF174603.1 | ||||
L24178.1 | AAA19560.1 | |||
M95167.1 | AAC41720.1 | |||
S44626.1 | AAB23443.1 | |||
S46955.1 | AAA11754.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv950628 | Chr.5:1380586 - 1414985 on Build GRCh38 | Deletion |
![]() |
esv3603807 | Chr.5:1384232 - 1416519 on Build GRCh38 | Loss |
![]() |
nsv830187 | Chr.5:1317991 - 1509711 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv596629 | Chr.5:457146 - 1468172 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv509045 | Chr.5:1282173 - 1455603 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)