Assay Details
Target Gene Details
Entrez Gene ID: | 1635 |
Gene Name: | dCMP deaminase |
Gene Aliases: |
- |
Location: |
Chr.4:182890091-182917477 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1016376 | Chr.4:182859067 - 183262599 on Build GRCh38 | Gain | WWC2-AS2 FAM92A1P2 WWC2-AS1 WWC2 DCTD |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |